Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013839.3 | NP_001013861.1 | 6 - 7 | 900 | 86 |
NM_001145297.3 | NP_001138769.1 | 6 - 7 | 900 | 86 | |
NM_001145299.3 | NP_001138771.1 | 6 - 7 | 900 | 86 | |
NM_001282313.1 | NP_001269242.1 | 5 - 6 | 890 | 86 | |
XM_006721786.3 | XP_006721849.1 | 6 - 7 | 899 | 86 | |
XM_006721787.3 | XP_006721850.1 | 6 - 7 | 1334 | 86 | |
XM_011524571.2 | XP_011522873.1 | 4 - 5 | 679 | 86 | |
XM_011524572.2 | XP_011522874.1 | 6 - 7 | 899 | 86 | |
GenBank mRNA | AB028990.1 | - | 6 - 7 | 823 | 86 |
AK295871.1 | - | 5 - 6 | 842 | 86 | |
AL834324.1 | - | 6 - 7 | 871 | 86 | |
AY869728.1 | - | 4 - 5 | 669 | 86 | |
BC011045.1 | - | 6 - 7 | 834 | 86 | |
BC018466.1 | - | 6 - 7 | 808 | 86 | |
FJ457120.1 | - | 6 - 7 | 805 | 86 |