Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005263458.3 | XP_005263515.1 | 1 - 2 | 1055 | 83 |
XM_011541496.2 | XP_011539798.1 | 1 - 2 | 1055 | 83 | |
XM_017001328.1 | XP_016856817.1 | 1 - 2 | 1055 | 83 | |
GenBank mRNA | AY046562.1 | - | 2 - 3 | 1164 | 83 |