Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004667.5 | NP_004658.3 | 79 - 80 | 12342 | 81 |
XM_005268276.4 | XP_005268333.1 | 78 - 79 | 12472 | 81 | |
XM_006720726.3 | XP_006720789.1 | 79 - 80 | 12348 | 81 | |
XM_006720727.3 | XP_006720790.1 | 77 - 78 | 12105 | 81 | |
XM_011522131.2 | XP_011520433.1 | 77 - 78 | 12129 | 81 | |
XM_011522133.2 | XP_011520435.1 | 60 - 61 | 9108 | 81 | |
XM_017022695.1 | XP_016878184.1 | 78 - 79 | 12289 | 81 | |
XM_017022696.1 | XP_016878185.1 | 78 - 79 | 12232 | 81 | |
XM_017022697.1 | XP_016878186.1 | 38 - 39 | 5671 | 81 | |
XM_017022698.1 | XP_016878187.1 | 38 - 39 | 5582 | 81 | |
GenBank mRNA | AF071172.3 | - | 79 - 80 | 12294 | 81 |
BX440995.2 | - | 5 - 6 | 721 | 81 | |
CN302846.1 | - | 3 - 4 | 480 | 81 |