Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004667.5 | NP_004658.3 | 85 - 86 | 13297 | 80 |
XM_005268276.4 | XP_005268333.1 | 84 - 85 | 13427 | 80 | |
XM_006720726.3 | XP_006720789.1 | 85 - 86 | 13303 | 80 | |
XM_006720727.3 | XP_006720790.1 | 83 - 84 | 13060 | 80 | |
XM_011522131.2 | XP_011520433.1 | 83 - 84 | 13084 | 80 | |
XM_011522133.2 | XP_011520435.1 | 66 - 67 | 10063 | 80 | |
XM_017022695.1 | XP_016878184.1 | 84 - 85 | 13244 | 80 | |
XM_017022696.1 | XP_016878185.1 | 84 - 85 | 13187 | 80 | |
XM_017022697.1 | XP_016878186.1 | 44 - 45 | 6626 | 80 | |
XM_017022698.1 | XP_016878187.1 | 44 - 45 | 6537 | 80 | |
GenBank mRNA | AF071172.3 | - | 85 - 86 | 13249 | 80 |