Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290263.1 | NP_001277192.1 | 39 - 40 | 4369 | 73 |
NM_014689.2 | NP_055504.2 | 39 - 40 | 4512 | 73 | |
XM_005246673.4 | XP_005246730.1 | 39 - 40 | 4371 | 73 | |
XM_006712617.3 | XP_006712680.1 | 39 - 40 | 4536 | 73 | |
XM_017004441.1 | XP_016859930.1 | 39 - 40 | 4539 | 73 | |
XM_017004442.1 | XP_016859931.1 | 39 - 40 | 4515 | 73 | |
XM_017004443.1 | XP_016859932.1 | 39 - 40 | 4588 | 73 | |
XM_017004444.1 | XP_016859933.1 | 38 - 39 | 4495 | 73 | |
XM_017004445.1 | XP_016859934.1 | 39 - 40 | 4369 | 73 | |
XM_017004446.1 | XP_016859935.1 | 38 - 39 | 4393 | 73 | |
XM_017004447.1 | XP_016859936.1 | 39 - 40 | 4511 | 73 | |
GenBank mRNA | AK293847.1 | - | 5 - 6 | 438 | 73 |
AK293879.1 | - | 10 - 11 | 918 | 73 | |
CR749492.1 | - | 14 - 15 | 1357 | 73 | |
EU236710.2 | - | 39 - 40 | 4366 | 73 |