Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290263.1 | NP_001277192.1 | 46 - 47 | 5311 | 80 |
NM_014689.2 | NP_055504.2 | 46 - 47 | 5454 | 80 | |
XM_005246673.4 | XP_005246730.1 | 46 - 47 | 5313 | 80 | |
XM_006712617.3 | XP_006712680.1 | 46 - 47 | 5478 | 80 | |
XM_017004441.1 | XP_016859930.1 | 46 - 47 | 5481 | 80 | |
XM_017004442.1 | XP_016859931.1 | 46 - 47 | 5457 | 80 | |
XM_017004443.1 | XP_016859932.1 | 46 - 47 | 5530 | 80 | |
XM_017004444.1 | XP_016859933.1 | 45 - 46 | 5437 | 80 | |
XM_017004445.1 | XP_016859934.1 | 46 - 47 | 5311 | 80 | |
XM_017004446.1 | XP_016859935.1 | 45 - 46 | 5335 | 80 | |
XM_017004447.1 | XP_016859936.1 | 46 - 47 | 5453 | 80 | |
GenBank mRNA | AK000227.1 | - | 4 - 5 | 423 | 80 |
AK001253.1 | - | 1 - 2 | 54 | 80 | |
AK293847.1 | - | 12 - 13 | 1380 | 80 | |
AK307914.1 | - | 6 - 7 | 815 | 80 | |
BC015018.1 | - | 4 - 5 | 421 | 80 | |
CR749492.1 | - | 21 - 22 | 2299 | 80 | |
EU236710.2 | - | 46 - 47 | 5308 | 80 |