Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007225.1 | NP_001007226.1 | 10 - 11 | 1210 | 104 |
NM_001291869.1 | NP_001278798.1 | 11 - 12 | 1357 | 104 | |
NM_001291872.1 | NP_001278801.1 | 10 - 11 | 1159 | 104 | |
NM_001291873.1 | NP_001278802.1 | 10 - 11 | 1141 | 104 | |
NM_001291874.1 | NP_001278803.1 | 9 - 10 | 1012 | 104 | |
NM_001291875.1 | NP_001278804.1 | 9 - 10 | 1077 | 104 | |
NM_006548.4 | NP_006539.3 | 11 - 12 | 1339 | 104 | |
XM_006713477.3 | XP_006713540.1 | 6 - 7 | 763 | 104 | |
XM_006713478.3 | XP_006713541.1 | 5 - 6 | 629 | 104 | |
XM_017005557.1 | XP_016861046.1 | 11 - 12 | 1336 | 104 | |
XM_017005558.1 | XP_016861047.1 | 10 - 11 | 1233 | 104 | |
XM_017005559.1 | XP_016861048.1 | 11 - 12 | 1363 | 104 | |
XM_017005560.1 | XP_016861049.1 | 11 - 12 | 1259 | 104 | |
XM_017005561.1 | XP_016861050.1 | 11 - 12 | 1364 | 104 | |
XM_017005562.1 | XP_016861051.1 | 11 - 12 | 1363 | 104 | |
GenBank mRNA | AF117107.1 | - | 11 - 12 | 1306 | 104 |
AK290664.1 | - | 11 - 12 | 1340 | 104 | |
AK296703.1 | - | 5 - 6 | 474 | 104 | |
BC021290.2 | - | 11 - 12 | 1324 | 104 | |
EU408701.1 | - | 10 - 11 | 1159 | 104 | |
EU408702.1 | - | 10 - 11 | 1141 | 104 | |
EU408703.1 | - | 9 - 10 | 1012 | 104 | |
EU408704.1 | - | 9 - 10 | 1077 | 104 |