Assay Design



Gene Transcripts

Gene Symbol
KALRN
Entrez Gene ID
8997
Gene Name
kalirin, RhoGEF kinase
Gene Aliases
ARHGEF24, CHD5, CHDS5, DUET, DUO, HAPIP, TRAD
Chromosome Location
Chr.3: 124033341 - 124726325 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024660.4 NP_001019831.2 8 - 9 1571 86
NM_001322988.1 NP_001309917.1 8 - 9 1571 86
NM_001322989.1 NP_001309918.1 8 - 9 1571 86
NM_001322990.1 NP_001309919.1 8 - 9 1571 86
NM_001322991.1 NP_001309920.1 8 - 9 1571 86
NM_001322992.1 NP_001309921.1 8 - 9 1571 86
NM_003947.5 NP_003938.1 8 - 9 1571 86
XM_006713810.3 XP_006713873.1 8 - 9 1816 86
XM_006713811.3 XP_006713874.1 8 - 9 1816 86
XM_006713812.3 XP_006713875.1 8 - 9 1816 86
XM_006713813.3 XP_006713876.1 8 - 9 1816 86
XM_006713814.3 XP_006713877.1 8 - 9 1816 86
XM_006713815.3 XP_006713878.1 8 - 9 1819 86
XM_011513279.2 XP_011511581.1 8 - 9 1816 86
XM_011513280.2 XP_011511582.1 8 - 9 1817 86
XM_011513281.2 XP_011511583.1 8 - 9 24465 86
XM_011513283.2 XP_011511585.1 8 - 9 1813 86
XM_011513285.2 XP_011511587.1 8 - 9 1812 86
XM_017007429.1 XP_016862918.1 8 - 9 1816 86
XM_017007430.1 XP_016862919.1 8 - 9 1816 86
XM_017007431.1 XP_016862920.1 8 - 9 1812 86
XM_017007432.1 XP_016862921.1 8 - 9 1812 86
XM_017007433.1 XP_016862922.1 8 - 9 1819 86
XM_017007434.1 XP_016862923.1 8 - 9 1819 86
XM_017007435.1 XP_016862924.1 8 - 9 1819 86
GenBank mRNA AK314126.1 - 8 - 9 1571 86
U94190.1 - 8 - 9 1514 86

View Full Product Details