Assay Design



Gene Transcripts

Gene Symbol
KALRN
Entrez Gene ID
8997
Gene Name
kalirin, RhoGEF kinase
Gene Aliases
ARHGEF24, CHD5, CHDS5, DUET, DUO, HAPIP, TRAD
Chromosome Location
Chr.3: 124033341 - 124726325 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024660.4 NP_001019831.2 40 - 41 6117 101
NM_001322988.1 NP_001309917.1 40 - 41 6114 101
NM_001322993.1 NP_001309922.1 7 - 8 1032 101
NM_001322994.1 NP_001309923.1 7 - 8 1029 101
NM_001322995.1 NP_001309924.1 7 - 8 1032 101
NM_001322996.1 NP_001309925.1 7 - 8 1032 101
NM_001322997.1 NP_001309926.1 7 - 8 1029 101
NM_001322998.1 NP_001309927.1 6 - 7 936 101
NM_001322999.1 NP_001309928.1 6 - 7 933 101
NM_001323000.1 NP_001309929.1 6 - 7 936 101
NM_001323001.1 NP_001309930.1 6 - 7 933 101
NM_007064.4 NP_008995.2 7 - 8 1032 101
XM_006713810.3 XP_006713873.1 40 - 41 6362 101
XM_006713811.3 XP_006713874.1 40 - 41 6335 101
XM_006713812.3 XP_006713875.1 40 - 41 6323 101
XM_006713813.3 XP_006713876.1 40 - 41 6296 101
XM_006713814.3 XP_006713877.1 39 - 40 6266 101
XM_011513279.2 XP_011511581.1 40 - 41 6362 101
XM_011513280.2 XP_011511582.1 40 - 41 6360 101
XM_011513281.2 XP_011511583.1 40 - 41 28984 101
XM_011513283.2 XP_011511585.1 40 - 41 6359 101
XM_011513285.2 XP_011511587.1 39 - 40 6262 101
XM_017007429.1 XP_016862918.1 40 - 41 6362 101
XM_017007430.1 XP_016862919.1 39 - 40 6263 101
XM_017007431.1 XP_016862920.1 40 - 41 6355 101
XM_017007432.1 XP_016862921.1 39 - 40 6259 101
GenBank mRNA AB011422.1 - 7 - 8 986 101
AK125979.1 - 7 - 8 1147 101
AK296657.1 - 4 - 5 440 101

View Full Product Details