Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170649.1 | NP_001164120.1 | 3 - 4 | 527 | 81 |
NM_001170650.1 | NP_001164121.1 | 2 - 3 | 401 | 81 | |
NM_031206.4 | NP_112483.1 | 3 - 4 | 527 | 81 | |
XM_005262301.1 | XP_005262358.1 | 3 - 4 | 512 | 81 | |
XM_005262304.1 | XP_005262361.1 | 3 - 4 | 512 | 81 | |
XM_005262305.3 | XP_005262362.1 | 3 - 4 | 512 | 81 | |
XM_005262306.3 | XP_005262363.1 | 3 - 4 | 512 | 81 | |
XM_011531045.1 | XP_011529347.1 | 2 - 3 | 376 | 81 | |
XM_011531046.1 | XP_011529348.1 | 3 - 4 | 512 | 81 | |
XM_017029877.1 | XP_016885366.1 | 3 - 4 | 512 | 81 | |
XM_017029878.1 | XP_016885367.1 | 3 - 4 | 512 | 81 | |
XM_017029879.1 | XP_016885368.1 | 3 - 4 | 512 | 81 | |
XM_017029880.1 | XP_016885369.1 | 3 - 4 | 512 | 81 | |
GenBank mRNA | AF116730.1 | - | 3 - 4 | 465 | 81 |
AK022587.1 | - | 3 - 4 | 477 | 81 | |
AK074087.1 | - | 3 - 4 | 496 | 81 | |
AK097383.1 | - | 2 - 3 | 351 | 81 | |
AK225059.1 | - | 3 - 4 | 460 | 81 | |
BC014545.1 | - | 3 - 4 | 511 | 81 | |
BC018610.1 | - | 3 - 4 | 475 | 81 | |
BC019302.1 | - | 3 - 4 | 472 | 81 | |
DA619280.1 | - | 3 - 4 | 528 | 81 |