Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001290.1 | NP_001001290.1 | 4 - 5 | 488 | 105 |
NM_020041.2 | NP_064425.2 | 3 - 4 | 477 | 105 | |
XM_006713968.3 | XP_006714031.1 | 3 - 4 | 589 | 105 | |
XM_006713969.2 | XP_006714032.1 | 4 - 5 | 450 | 105 | |
XM_011513856.2 | XP_011512158.1 | 3 - 4 | 614 | 105 | |
XM_011513857.1 | XP_011512159.1 | 4 - 5 | 450 | 105 | |
XM_011513858.1 | XP_011512160.1 | 4 - 5 | 652 | 105 | |
XM_011513859.2 | XP_011512161.1 | 3 - 4 | 584 | 105 | |
XM_011513860.2 | XP_011512162.1 | 3 - 4 | 584 | 105 | |
XM_011513861.2 | XP_011512163.1 | 3 - 4 | 586 | 105 | |
XM_011513865.2 | XP_011512167.1 | 3 - 4 | 588 | 105 | |
XM_011513866.2 | XP_011512168.1 | 3 - 4 | 586 | 105 | |
XM_011513868.2 | XP_011512170.1 | 3 - 4 | 605 | 105 | |
XM_017008457.1 | XP_016863946.1 | 3 - 4 | 602 | 105 | |
XM_017008458.1 | XP_016863947.1 | 3 - 4 | 587 | 105 | |
GenBank mRNA | AF210317.1 | - | 3 - 4 | 470 | 105 |
AF421859.1 | - | 1 - 2 | 165 | 105 | |
AK310895.1 | - | 4 - 5 | 450 | 105 | |
BC018897.2 | - | 4 - 5 | 488 | 105 | |
BC110414.1 | - | 3 - 4 | 507 | 105 | |
CD629819.1 | - | 3 - 4 | 482 | 105 | |
CD629822.1 | - | 3 - 4 | 105 |