Assay Design



Gene Transcripts

Gene Symbol
SLC2A9
Entrez Gene ID
56606
Gene Name
solute carrier family 2 member 9
Gene Aliases
GLUT9, GLUTX, UAQTL2, URATv1
Chromosome Location
Chr.4: 9771125 - 10040248 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001290.1 NP_001001290.1 6 - 7 755 80
NM_020041.2 NP_064425.2 5 - 6 744 80
XM_006713968.3 XP_006714031.1 5 - 6 856 80
XM_006713969.2 XP_006714032.1 6 - 7 717 80
XM_011513856.2 XP_011512158.1 5 - 6 881 80
XM_011513857.1 XP_011512159.1 6 - 7 717 80
XM_011513858.1 XP_011512160.1 6 - 7 919 80
XM_011513859.2 XP_011512161.1 5 - 6 851 80
XM_011513860.2 XP_011512162.1 5 - 6 851 80
XM_011513861.2 XP_011512163.1 5 - 6 853 80
XM_011513862.2 XP_011512164.1 4 - 5 728 80
XM_011513863.2 XP_011512165.1 4 - 5 2407 80
XM_011513864.2 XP_011512166.1 3 - 4 296 80
XM_011513865.2 XP_011512167.1 5 - 6 855 80
XM_011513866.2 XP_011512168.1 5 - 6 853 80
XM_011513867.2 XP_011512169.1 2 - 3 810 80
XM_011513868.2 XP_011512170.1 5 - 6 872 80
XM_017008457.1 XP_016863946.1 5 - 6 869 80
XM_017008458.1 XP_016863947.1 5 - 6 854 80
XM_017008459.1 XP_016863948.1 3 - 4 388 80
XM_017008460.1 XP_016863949.1 4 - 5 2401 80
GenBank mRNA AF210317.1 - 5 - 6 737 80
AF421859.1 - 3 - 4 432 80
AK310895.1 - 6 - 7 717 80
BC018897.2 - 6 - 7 755 80
BC110414.1 - 5 - 6 774 80
CD629822.1 - 1 - 2 80

View Full Product Details