Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031280.3 | NP_112570.2 | 2 - 3 | 337 | 95 |
GenBank mRNA | AB049946.1 | - | 2 - 3 | 212 | 95 |
AB209905.1 | - | 2 - 3 | 236 | 95 | |
AK302633.1 | - | 2 - 3 | 281 | 95 | |
AK315441.1 | - | 2 - 3 | 276 | 95 | |
BC031336.1 | - | 2 - 3 | 252 | 95 | |
BM048518.1 | - | 2 - 3 | 336 | 95 | |
BU927793.1 | - | 2 - 3 | 187 | 95 |