Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001141973.2 | NP_001135445.1 | 17 - 18 | 2062 | 115 |
NM_001141974.2 | NP_001135446.1 | 17 - 18 | 2062 | 115 | |
NM_022089.3 | NP_071372.1 | 17 - 18 | 2077 | 115 | |
XM_005245810.1 | XP_005245867.1 | 17 - 18 | 2074 | 115 | |
XM_005245811.1 | XP_005245868.1 | 17 - 18 | 2062 | 115 | |
XM_005245812.1 | XP_005245869.1 | 17 - 18 | 2050 | 115 | |
XM_005245815.1 | XP_005245872.1 | 17 - 18 | 2077 | 115 | |
XM_006710512.1 | XP_006710575.1 | 17 - 18 | 2059 | 115 | |
XM_006710513.1 | XP_006710576.1 | 17 - 18 | 2035 | 115 | |
XM_011541128.1 | XP_011539430.1 | 17 - 18 | 2062 | 115 | |
XM_011541129.1 | XP_011539431.1 | 16 - 17 | 1870 | 115 | |
XM_017000844.1 | XP_016856333.1 | 17 - 18 | 2062 | 115 | |
XM_017000845.1 | XP_016856334.1 | 17 - 18 | 2059 | 115 | |
XM_017000846.1 | XP_016856335.1 | 17 - 18 | 2035 | 115 | |
XM_017000847.1 | XP_016856336.1 | 17 - 18 | 2032 | 115 | |
XM_017000848.1 | XP_016856337.1 | 17 - 18 | 2077 | 115 | |
XM_017000849.1 | XP_016856338.1 | 17 - 18 | 2062 | 115 | |
XM_017000850.1 | XP_016856339.1 | 16 - 17 | 1870 | 115 | |
GenBank mRNA | AK075310.1 | - | 7 - 8 | 859 | 115 |
AK290210.1 | - | 17 - 18 | 2018 | 115 | |
AL354615.1 | - | 17 - 18 | 1877 | 115 | |
AY461712.1 | - | 17 - 18 | 1876 | 115 | |
BC030267.1 | - | 17 - 18 | 2005 | 115 | |
BC034575.1 | - | 14 - 15 | 1568 | 115 |