Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001141973.2 | NP_001135445.1 | 21 - 22 | 2630 | 79 |
NM_022089.3 | NP_071372.1 | 21 - 22 | 2645 | 79 | |
XM_005245810.1 | XP_005245867.1 | 21 - 22 | 2642 | 79 | |
XM_005245811.1 | XP_005245868.1 | 21 - 22 | 2630 | 79 | |
XM_005245812.1 | XP_005245869.1 | 21 - 22 | 2618 | 79 | |
XM_006710512.1 | XP_006710575.1 | 21 - 22 | 2627 | 79 | |
XM_006710513.1 | XP_006710576.1 | 21 - 22 | 2603 | 79 | |
XM_011541128.1 | XP_011539430.1 | 21 - 22 | 2630 | 79 | |
XM_011541129.1 | XP_011539431.1 | 20 - 21 | 2438 | 79 | |
XM_017000844.1 | XP_016856333.1 | 21 - 22 | 2630 | 79 | |
XM_017000845.1 | XP_016856334.1 | 21 - 22 | 2627 | 79 | |
XM_017000846.1 | XP_016856335.1 | 21 - 22 | 2603 | 79 | |
XM_017000847.1 | XP_016856336.1 | 21 - 22 | 2600 | 79 | |
XM_017000850.1 | XP_016856339.1 | 20 - 21 | 2438 | 79 | |
GenBank mRNA | AK075310.1 | - | 11 - 12 | 1427 | 79 |
AK290210.1 | - | 21 - 22 | 2586 | 79 | |
AL354615.1 | - | 21 - 22 | 2445 | 79 | |
AL833966.1 | - | 2 - 3 | 299 | 79 | |
AY461712.1 | - | 21 - 22 | 2444 | 79 | |
BC034575.1 | - | 18 - 19 | 2136 | 79 |