Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001141973.2 | NP_001135445.1 | 27 - 28 | 3460 | 130 |
NM_022089.3 | NP_071372.1 | 27 - 28 | 3475 | 130 | |
XM_017000844.1 | XP_016856333.1 | 27 - 28 | 3460 | 130 | |
XM_017000845.1 | XP_016856334.1 | 27 - 28 | 3457 | 130 | |
XM_017000846.1 | XP_016856335.1 | 27 - 28 | 3433 | 130 | |
XM_017000847.1 | XP_016856336.1 | 27 - 28 | 3430 | 130 | |
XM_017000848.1 | XP_016856337.1 | 26 - 27 | 3358 | 130 | |
XM_017000849.1 | XP_016856338.1 | 26 - 27 | 3343 | 130 | |
XM_017000850.1 | XP_016856339.1 | 26 - 27 | 3268 | 130 | |
GenBank mRNA | AK025928.1 | - | 7 - 8 | 864 | 130 |
AK075310.1 | - | 17 - 18 | 2257 | 130 | |
AK290210.1 | - | 27 - 28 | 3416 | 130 | |
AL354615.1 | - | 27 - 28 | 3275 | 130 | |
AY461712.1 | - | 27 - 28 | 3274 | 130 | |
BC034575.1 | - | 24 - 25 | 2966 | 130 |