Assay Design



Gene Transcripts

Gene Symbol
RCC2
Entrez Gene ID
55920
Gene Name
regulator of chromosome condensation 2
Gene Aliases
TD-60
Chromosome Location
Chr.1: 17406755 - 17439754 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018715.3 NP_061185.1 1 - 2 216 88
GenBank mRNA AB040903.1 - 1 - 2 147 88
AJ421269.1 - 1 - 2 233 88
DR000303.1 - 1 - 2 222 88

View Full Product Details