Assay Design



Gene Transcripts

Gene Symbol
KCNMA1
Entrez Gene ID
3778
Gene Name
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
Chromosome Location
Chr.10: 76869601 - 77637819 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014797.2 NP_001014797.1 11 - 12 1620 84
NM_001161352.1 NP_001154824.1 11 - 12 1620 84
NM_001161353.1 NP_001154825.1 11 - 12 1620 84
NM_001271518.1 NP_001258447.1 10 - 11 1458 84
NM_001271519.1 NP_001258448.1 11 - 12 1620 84
NM_001322829.1 NP_001309758.1 11 - 12 1620 84
NM_001322830.1 NP_001309759.1 11 - 12 1620 84
NM_001322832.1 NP_001309761.1 11 - 12 1620 84
NM_001322835.1 NP_001309764.1 11 - 12 1620 84
NM_001322836.1 NP_001309765.1 11 - 12 1620 84
NM_001322837.1 NP_001309766.1 11 - 12 1620 84
NM_001322838.1 NP_001309767.1 12 - 13 1580 84
NM_002247.3 NP_002238.2 11 - 12 1620 84
XM_005269776.3 XP_005269833.1 11 - 12 1609 84
XM_005269778.1 XP_005269835.1 11 - 12 1609 84
XM_005269781.2 XP_005269838.1 11 - 12 1541 84
XM_005269787.3 XP_005269844.1 11 - 12 1609 84
XM_005269789.2 XP_005269846.1 11 - 12 1541 84
XM_005269792.2 XP_005269849.1 11 - 12 1541 84
XM_005269796.2 XP_005269853.1 11 - 12 1541 84
XM_006717826.2 XP_006717889.1 11 - 12 1541 84
XM_011539773.1 XP_011538075.1 12 - 13 1741 84
XM_011539774.1 XP_011538076.1 12 - 13 1741 84
XM_011539775.1 XP_011538077.1 12 - 13 1741 84
XM_011539776.1 XP_011538078.1 12 - 13 1741 84
XM_011539777.1 XP_011538079.1 11 - 12 1609 84
XM_011539778.2 XP_011538080.1 11 - 12 1541 84
XM_011539779.1 XP_011538081.1 12 - 13 1741 84
XM_011539780.1 XP_011538082.1 11 - 12 1609 84
XM_011539781.2 XP_011538083.1 12 - 13 1741 84
XM_011539782.2 XP_011538084.1 12 - 13 1693 84
XM_011539783.1 XP_011538085.1 11 - 12 1609 84
XM_011539784.1 XP_011538086.1 12 - 13 1741 84
XM_011539785.1 XP_011538087.1 11 - 12 1609 84
XM_017016207.1 XP_016871696.1 12 - 13 1741 84
XM_017016208.1 XP_016871697.1 12 - 13 1741 84
XM_017016209.1 XP_016871698.1 12 - 13 1741 84
XM_017016210.1 XP_016871699.1 12 - 13 1741 84
XM_017016211.1 XP_016871700.1 12 - 13 1741 84
XM_017016212.1 XP_016871701.1 12 - 13 1741 84
XM_017016213.1 XP_016871702.1 12 - 13 1741 84
XM_017016214.1 XP_016871703.1 12 - 13 1741 84
XM_017016215.1 XP_016871704.1 11 - 12 1609 84
XM_017016216.1 XP_016871705.1 12 - 13 1741 84
XM_017016217.1 XP_016871706.1 12 - 13 1693 84
XM_017016218.1 XP_016871707.1 10 - 11 1066 84
XM_017016219.1 XP_016871708.1 12 - 13 1741 84
XM_017016222.1 XP_016871711.1 11 - 12 1609 84
XM_017016223.1 XP_016871712.1 9 - 10 918 84
GenBank mRNA AB113382.1 - 11 - 12 1385 84
AB113575.1 - 11 - 12 1371 84
AB209488.1 - 10 - 11 1026 84
AB524033.1 - 10 - 11 1281 84
AF025999.1 - 11 - 12 1371 84
AF118141.1 - 11 - 12 1248 84
AK310379.1 - 12 - 13 1580 84
AY040849.1 - 11 - 12 1385 84
BC137115.1 - 11 - 12 1523 84
BC137137.1 - 11 - 12 1523 84
BC144496.1 - 11 - 12 1523 84
BX648925.1 - 6 - 7 602 84
U02632.1 - 4 - 5 478 84
U09384.1 - 11 - 12 1438 84
U11058.2 - 11 - 12 1334 84
U11717.1 - 11 - 12 1395 84
U13913.1 - 11 - 12 1521 84
U23767.1 - 11 - 12 1443 84

View Full Product Details