Assay Design



Gene Transcripts

Gene Symbol
KCNMA1
Entrez Gene ID
3778
Gene Name
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
Chromosome Location
Chr.10: 76869601 - 77637819 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014797.2 NP_001014797.1 24 - 25 3031 98
NM_001161352.1 NP_001154824.1 24 - 25 3193 98
NM_001161353.1 NP_001154825.1 24 - 25 3142 98
NM_001271518.1 NP_001258447.1 23 - 24 2869 98
NM_001322829.1 NP_001309758.1 24 - 25 3028 98
NM_001322832.1 NP_001309761.1 23 - 24 3019 98
NM_001322836.1 NP_001309765.1 24 - 25 3028 98
NM_001322838.1 NP_001309767.1 25 - 26 3066 98
NM_002247.3 NP_002238.2 23 - 24 3019 98
XM_005269778.1 XP_005269835.1 25 - 26 3194 98
XM_005269781.2 XP_005269838.1 24 - 25 3114 98
XM_005269787.3 XP_005269844.1 23 - 24 3008 98
XM_005269792.2 XP_005269849.1 24 - 25 3027 98
XM_005269796.2 XP_005269853.1 23 - 24 2940 98
XM_011539773.1 XP_011538075.1 26 - 27 3326 98
XM_011539775.1 XP_011538077.1 25 - 26 3314 98
XM_011539776.1 XP_011538078.1 25 - 26 3314 98
XM_011539779.1 XP_011538081.1 25 - 26 3227 98
XM_011539780.1 XP_011538082.1 25 - 26 3194 98
XM_011539781.2 XP_011538083.1 25 - 26 3152 98
XM_011539782.2 XP_011538084.1 24 - 25 3092 98
XM_011539783.1 XP_011538085.1 25 - 26 3107 98
XM_017016210.1 XP_016871699.1 26 - 27 3326 98
XM_017016211.1 XP_016871700.1 26 - 27 3239 98
XM_017016213.1 XP_016871702.1 25 - 26 3227 98
XM_017016216.1 XP_016871705.1 25 - 26 3152 98
XM_017016217.1 XP_016871706.1 24 - 25 3092 98
XM_017016222.1 XP_016871711.1 23 - 24 3008 98
XM_017016223.1 XP_016871712.1 22 - 23 2404 98
GenBank mRNA AB113382.1 - 24 - 25 2958 98
AB113575.1 - 24 - 25 2893 98
AB209488.1 - 23 - 24 2438 98
AB524033.1 - 23 - 24 2692 98
AF025999.1 - 23 - 24 2770 98
AF118141.1 - 23 - 24 2647 98
AK310379.1 - 25 - 26 3066 98
AY040849.1 - 23 - 24 2784 98
BC137115.1 - 23 - 24 2922 98
BC137137.1 - 23 - 24 2922 98
BX648925.1 - 20 - 21 2184 98
U02632.1 - 17 - 18 1964 98
U11058.2 - 23 - 24 2733 98
U11717.1 - 23 - 24 2794 98
U13913.1 - 23 - 24 2920 98
U23767.1 - 23 - 24 2842 98

View Full Product Details