Assay Design



Gene Transcripts

Gene Symbol
ANO3
Entrez Gene ID
63982
Gene Name
anoctamin 3
Gene Aliases
C11orf25, DYT23, DYT24, GENX-3947, TMEM16C
Chromosome Location
Chr.11: 26189123 - 26663289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313726.1 NP_001300655.1 11 - 12 1271 65
NM_001313727.1 NP_001300656.1 7 - 8 829 65
NM_031418.3 NP_113606.2 10 - 11 1179 65
XM_017018118.1 XP_016873607.1 8 - 9 807 65
XM_017018119.1 XP_016873608.1 5 - 6 419 65
GenBank mRNA AJ300461.1 - 10 - 11 1886 65
AK295143.1 - 7 - 8 742 65
AK295816.1 - 7 - 8 741 65
AK302659.1 - 10 - 11 1218 65
AK304781.1 - 11 - 12 1112 65
AK316018.1 - 7 - 8 741 65

View Full Product Details