Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001313726.1 | NP_001300655.1 | 27 - 28 | 2998 | 79 |
NM_001313727.1 | NP_001300656.1 | 23 - 24 | 2556 | 79 | |
NM_031418.3 | NP_113606.2 | 26 - 27 | 2906 | 79 | |
XM_011520282.2 | XP_011518584.1 | 13 - 14 | 1453 | 79 | |
XM_017018118.1 | XP_016873607.1 | 24 - 25 | 2534 | 79 | |
XM_017018119.1 | XP_016873608.1 | 21 - 22 | 2146 | 79 | |
GenBank mRNA | AJ300461.1 | - | 26 - 27 | 3613 | 79 |
AK295658.1 | - | 10 - 11 | 941 | 79 | |
AK295816.1 | - | 23 - 24 | 2468 | 79 | |
AK302659.1 | - | 26 - 27 | 2945 | 79 | |
AK304781.1 | - | 27 - 28 | 2839 | 79 | |
AK316018.1 | - | 23 - 24 | 2468 | 79 |