Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032172.2 | NP_115548.1 | 12 - 13 | 1510 | 73 |
XM_005249883.4 | XP_005249940.1 | 12 - 13 | 1457 | 73 | |
XM_006715790.1 | XP_006715853.1 | 12 - 13 | 1457 | 73 | |
XM_006715791.2 | XP_006715854.1 | 15 - 16 | 1963 | 73 | |
XM_011515573.1 | XP_011513875.1 | 16 - 17 | 2022 | 73 | |
XM_011515574.1 | XP_011513876.1 | 13 - 14 | 1546 | 73 | |
XM_011515577.1 | XP_011513879.1 | 11 - 12 | 1175 | 73 | |
XM_011515578.1 | XP_011513880.1 | 13 - 14 | 1597 | 73 | |
GenBank mRNA | AJ601395.1 | - | 11 - 12 | 1387 | 73 |
AK022759.1 | - | 12 - 13 | 1454 | 73 | |
AK301527.1 | - | 10 - 11 | 982 | 73 | |
AK309375.1 | - | 11 - 12 | 1251 | 73 | |
AY618868.1 | - | 12 - 13 | 1454 | 73 | |
BC060846.1 | - | 12 - 13 | 1449 | 73 | |
BC132862.1 | - | 11 - 12 | 1407 | 73 | |
BC136656.1 | - | 10 - 11 | 2340 | 73 |