Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032172.2 | NP_115548.1 | 2 - 3 | 363 | 166 |
XM_005249883.4 | XP_005249940.1 | 2 - 3 | 310 | 166 | |
XM_006715790.1 | XP_006715853.1 | 2 - 3 | 310 | 166 | |
XM_006715791.2 | XP_006715854.1 | 5 - 6 | 816 | 166 | |
XM_011515573.1 | XP_011513875.1 | 6 - 7 | 875 | 166 | |
XM_011515574.1 | XP_011513876.1 | 2 - 3 | 308 | 166 | |
XM_011515578.1 | XP_011513880.1 | 2 - 3 | 305 | 166 | |
GenBank mRNA | AJ601395.1 | - | 1 - 2 | 240 | 166 |
AK022759.1 | - | 2 - 3 | 307 | 166 | |
AK298164.1 | - | 2 - 3 | 334 | 166 | |
AY618868.1 | - | 2 - 3 | 307 | 166 | |
BC060846.1 | - | 2 - 3 | 302 | 166 | |
BC132862.1 | - | 1 - 2 | 260 | 166 | |
BC136656.1 | - | 1 - 2 | 260 | 166 | |
DB457216.2 | - | 2 - 3 | 166 | ||
DR001777.1 | - | 2 - 3 | 367 | 166 |