Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244871.1 | NP_001231800.1 | 2 - 3 | 621 | 78 |
NM_001343.3 | NP_001334.2 | 2 - 3 | 621 | 78 | |
GenBank mRNA | AF188298.1 | - | 1 - 2 | 178 | 78 |
AK314381.1 | - | 2 - 3 | 369 | 78 | |
BC003064.2 | - | 2 - 3 | 217 | 78 | |
U39050.1 | - | 1 - 2 | 189 | 78 |