Assay Design
Gene Transcripts
- Gene Symbol
- SVEP1
- Entrez Gene ID
-
79987
- Gene Name
- sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1
- Gene Aliases
-
C9orf13, CCP22, POLYDOM, SEL-OB, SELOB
- Chromosome Location
-
Chr.9: 110365248 - 110579880 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_153366.3 |
NP_699197.3 |
32 - 33 |
5693 |
63 |
GenBank mRNA |
DV080389.1 |
- |
1 - 2 |
117 |
63 |
View Full Product Details