Assay Design



Gene Transcripts

Gene Symbol
SVEP1
Entrez Gene ID
79987
Gene Name
sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1
Gene Aliases
C9orf13, CCP22, POLYDOM, SEL-OB, SELOB
Chromosome Location
Chr.9: 110365248 - 110579880 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153366.3 NP_699197.3 42 - 43 10398 86
GenBank mRNA AJ619977.1 - 42 - 43 10233 86
AK075234.1 - 5 - 6 3076 86
AL832416.1 - 5 - 6 2885 86
AY243503.1 - 42 - 43 10233 86
AY916667.1 - 6 - 7 3839 86

View Full Product Details