Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144030.1 | NP_001137502.1 | 17 - 18 | 2062 | 110 |
NM_024662.2 | NP_078938.2 | 19 - 20 | 2277 | 110 | |
GenBank mRNA | AB051496.1 | - | 19 - 20 | 2198 | 110 |
AF489535.1 | - | 15 - 16 | 1574 | 110 | |
AK001636.1 | - | 19 - 20 | 2132 | 110 | |
AK022241.1 | - | 19 - 20 | 2239 | 110 | |
AK074430.1 | - | - | 305 | 110 | |
AK225341.1 | - | 19 - 20 | 2239 | 110 | |
AK294044.1 | - | 17 - 18 | 1961 | 110 | |
AL136882.1 | - | 19 - 20 | 2134 | 110 | |
BC035558.1 | - | 19 - 20 | 2211 | 110 |