Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204824.1 | NP_001191753.1 | 13 - 14 | 1480 | 60 |
NM_004519.3 | NP_004510.1 | 13 - 14 | 2029 | 60 | |
XM_005250914.3 | XP_005250971.1 | 10 - 11 | 1104 | 60 | |
XM_006716555.3 | XP_006716618.1 | 11 - 12 | 1290 | 60 | |
XM_011517026.2 | XP_011515328.1 | 13 - 14 | 4563 | 60 | |
XM_017013400.1 | XP_016868889.1 | 13 - 14 | 11535 | 60 | |
GenBank mRNA | AB208890.1 | - | 12 - 13 | 2532 | 60 |
AF033347.1 | - | 13 - 14 | 1663 | 60 | |
AK296293.1 | - | 13 - 14 | 1480 | 60 |