Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204824.1 | NP_001191753.1 | 2 - 3 | 153 | 87 |
NM_004519.3 | NP_004510.1 | 2 - 3 | 702 | 87 | |
XM_011517026.2 | XP_011515328.1 | 2 - 3 | 3236 | 87 | |
XM_017013400.1 | XP_016868889.1 | 2 - 3 | 10208 | 87 | |
GenBank mRNA | AB208890.1 | - | 2 - 3 | 255 | 87 |
AF033347.1 | - | 2 - 3 | 336 | 87 | |
AK296293.1 | - | 2 - 3 | 153 | 87 | |
BC128576.1 | - | 3 - 4 | 445 | 87 |