Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204824.1 | NP_001191753.1 | 4 - 5 | 451 | 55 |
NM_004519.3 | NP_004510.1 | 4 - 5 | 1000 | 55 | |
XM_005250914.3 | XP_005250971.1 | 1 - 2 | 164 | 55 | |
XM_006716555.3 | XP_006716618.1 | 2 - 3 | 261 | 55 | |
XM_011517026.2 | XP_011515328.1 | 4 - 5 | 3534 | 55 | |
XM_017013400.1 | XP_016868889.1 | 4 - 5 | 10506 | 55 | |
GenBank mRNA | AF033347.1 | - | 4 - 5 | 634 | 55 |
AK296293.1 | - | 4 - 5 | 451 | 55 | |
BC128576.1 | - | 5 - 6 | 743 | 55 |