Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282659.1 | NP_001269588.1 | 8 - 9 | 1726 | 107 |
NM_017944.3 | NP_060414.3 | 7 - 8 | 1522 | 107 | |
XM_005252997.1 | XP_005253054.1 | 9 - 10 | 1786 | 107 | |
XM_017017953.1 | XP_016873442.1 | 9 - 10 | 1842 | 107 | |
GenBank mRNA | AK027283.1 | - | - | 634 | 107 |
AK027362.1 | - | 7 - 8 | 927 | 107 | |
AK092290.1 | - | 9 - 10 | 1143 | 107 | |
AK309240.1 | - | 8 - 9 | 1190 | 107 |