Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003017.4 | NP_003008.1 | 3 - 4 | 513 | 81 |
GenBank mRNA | AB451229.1 | - | 2 - 3 | 342 | 81 |
AB451352.1 | - | 2 - 3 | 342 | 81 | |
AF107405.1 | - | 3 - 4 | 437 | 81 | |
AK312553.1 | - | 3 - 4 | 464 | 81 | |
BC000914.1 | - | 3 - 4 | 447 | 81 | |
BC069018.1 | - | 3 - 4 | 443 | 81 | |
BT007017.1 | - | 2 - 3 | 342 | 81 | |
L10838.1 | - | 2 - 3 | 342 | 81 |