Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008938.3 | NP_001008938.1 | 11 - 12 | 1459 | 85 |
NM_014756.3 | NP_055571.2 | 11 - 12 | 1459 | 85 | |
GenBank mRNA | AW504660.1 | - | 1 - 2 | 98 | 85 |
BC111043.1 | - | 11 - 12 | 1411 | 85 | |
BC120869.1 | - | 10 - 11 | 1353 | 85 | |
BC120870.1 | - | 10 - 11 | 1351 | 85 | |
D43948.1 | - | 10 - 11 | 1380 | 85 | |
X92474.1 | - | 10 - 11 | 1359 | 85 |