Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008938.3 | NP_001008938.1 | 1 - 2 | 92 | 66 |
NM_014756.3 | NP_055571.2 | 1 - 2 | 92 | 66 | |
GenBank mRNA | BC017856.1 | - | 1 - 2 | 62 | 66 |
BC070136.1 | - | 1 - 2 | 62 | 66 | |
BC111043.1 | - | 1 - 2 | 44 | 66 | |
BP282525.1 | - | 1 - 2 | 92 | 66 |