Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008938.3 | NP_001008938.1 | 21 - 22 | 2697 | 124 |
NM_014756.3 | NP_055571.2 | 21 - 22 | 2697 | 124 | |
GenBank mRNA | BC120869.1 | - | 20 - 21 | 2591 | 124 |
BC120870.1 | - | 20 - 21 | 2589 | 124 | |
D43948.1 | - | 20 - 21 | 2618 | 124 | |
X92474.1 | - | 20 - 21 | 2597 | 124 |