Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005215.3 | NP_005206.2 | 22 - 23 | 3844 | 110 |
XM_011525843.1 | XP_011524145.1 | 22 - 23 | 3844 | 110 | |
XM_011525844.2 | XP_011524146.1 | 19 - 20 | 2732 | 110 | |
XM_017025568.1 | XP_016881057.1 | 22 - 23 | 3844 | 110 | |
XM_017025569.1 | XP_016881058.1 | 22 - 23 | 3784 | 110 | |
XM_017025570.1 | XP_016881059.1 | 18 - 19 | 2740 | 110 | |
GenBank mRNA | AK310385.1 | - | 19 - 20 | 2398 | 110 |
X76132.1 | - | 22 - 23 | 3228 | 110 |