Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005215.3 | NP_005206.2 | 9 - 10 | 2189 | 71 |
XM_011525843.1 | XP_011524145.1 | 9 - 10 | 2189 | 71 | |
XM_011525844.2 | XP_011524146.1 | 6 - 7 | 1077 | 71 | |
XM_017025568.1 | XP_016881057.1 | 9 - 10 | 2189 | 71 | |
XM_017025569.1 | XP_016881058.1 | 9 - 10 | 2189 | 71 | |
XM_017025570.1 | XP_016881059.1 | 5 - 6 | 1085 | 71 | |
GenBank mRNA | AK302643.1 | - | 9 - 10 | 1494 | 71 |
AK310385.1 | - | 6 - 7 | 803 | 71 | |
BC036524.1 | - | 8 - 9 | 1375 | 71 | |
M32292.1 | - | 9 - 10 | 1579 | 71 | |
X76132.1 | - | 9 - 10 | 1573 | 71 |