Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005215.3 | NP_005206.2 | 10 - 11 | 2334 | 152 |
XM_011525843.1 | XP_011524145.1 | 10 - 11 | 2334 | 152 | |
XM_011525844.2 | XP_011524146.1 | 7 - 8 | 1222 | 152 | |
XM_017025568.1 | XP_016881057.1 | 10 - 11 | 2334 | 152 | |
XM_017025569.1 | XP_016881058.1 | 10 - 11 | 2334 | 152 | |
XM_017025570.1 | XP_016881059.1 | 6 - 7 | 1230 | 152 | |
GenBank mRNA | AK302643.1 | - | 10 - 11 | 1639 | 152 |
AK310385.1 | - | 7 - 8 | 948 | 152 | |
BC036524.1 | - | 9 - 10 | 1520 | 152 | |
M32292.1 | - | 10 - 11 | 1724 | 152 | |
X76132.1 | - | 10 - 11 | 1718 | 152 |