Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014839.1 | NP_001014839.1 | 7 - 8 | 1879 | 57 |
NM_001014841.1 | NP_001014841.1 | 6 - 7 | 1811 | 57 | |
NM_014284.2 | NP_055099.1 | 6 - 7 | 2151 | 57 | |
GenBank mRNA | AB011179.1 | - | 6 - 7 | 1759 | 57 |
AK295979.1 | - | 2 - 3 | 335 | 57 | |
BC024592.1 | - | 6 - 7 | 2125 | 57 |