Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198593.1 | NP_001185522.1 | 3 - 4 | 2249 | 143 |
NM_001198594.1 | NP_001185523.1 | 2 - 3 | 2185 | 143 | |
NM_172311.2 | NP_758515.1 | 3 - 4 | 2252 | 143 | |
GenBank mRNA | AF026169.1 | - | 3 - 4 | 2252 | 143 |
AK309740.1 | - | 3 - 4 | 2249 | 143 | |
BC101772.1 | - | 2 - 3 | 2165 | 143 | |
BC101774.1 | - | 2 - 3 | 2165 | 143 | |
BC143520.1 | - | 2 - 3 | 2165 | 143 | |
BX091338.1 | - | 2 - 3 | 319 | 143 |