Assay Design



Gene Transcripts

Gene Symbol
KCNMA1
Entrez Gene ID
3778
Gene Name
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
Chromosome Location
Chr.10: 76869601 - 77637819 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322838.1 NP_001309767.1 19 - 20 2225 93
XM_005269792.2 XP_005269849.1 18 - 19 2186 93
XM_011539774.1 XP_011538076.1 20 - 21 2398 93
XM_011539779.1 XP_011538081.1 19 - 20 2386 93
XM_011539783.1 XP_011538085.1 19 - 20 2266 93
XM_017016211.1 XP_016871700.1 20 - 21 2398 93
XM_017016213.1 XP_016871702.1 19 - 20 2386 93
XM_017016215.1 XP_016871704.1 19 - 20 2266 93
XM_017016223.1 XP_016871712.1 16 - 17 1563 93
GenBank mRNA AK310379.1 - 19 - 20 2225 93
U02632.1 - 11 - 12 1123 93

View Full Product Details