Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307928.1 | NP_001294857.1 | 6 - 7 | 869 | 88 |
NM_080474.2 | NP_536722.1 | 6 - 7 | 809 | 88 | |
XM_005266778.3 | XP_005266835.2 | 7 - 8 | 908 | 88 | |
XM_011526246.1 | XP_011524548.1 | 8 - 9 | 1098 | 88 | |
XM_011526248.1 | XP_011524550.1 | 7 - 8 | 934 | 88 | |
XM_011526249.1 | XP_011524551.1 | 7 - 8 | 908 | 88 | |
XM_017026067.1 | XP_016881556.1 | 7 - 8 | 991 | 88 | |
GenBank mRNA | AF411191.1 | - | 6 - 7 | 809 | 88 |
AK309754.1 | - | 3 - 4 | 619 | 88 | |
BC103884.1 | - | 6 - 7 | 870 | 88 | |
BC103885.1 | - | 6 - 7 | 869 | 88 |