Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006350.3 | NP_006341.1 | 4 - 5 | 1075 | 142 |
NM_013409.2 | NP_037541.1 | 4 - 5 | 1075 | 142 | |
GenBank mRNA | AB451330.1 | - | 4 - 5 | 717 | 142 |
AB451474.1 | - | 4 - 5 | 717 | 142 | |
BC004107.2 | - | 4 - 5 | 922 | 142 | |
BQ949678.1 | - | 2 - 3 | 142 | ||
CR541813.1 | - | 4 - 5 | 717 | 142 |