Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014453.3 | NP_055268.1 | 1 - 2 | 140 | 66 |
XM_005258746.2 | XP_005258803.1 | 1 - 2 | 640 | 66 | |
GenBank mRNA | AF042384.1 | - | 1 - 2 | 108 | 66 |
BG546479.1 | - | 1 - 2 | 136 | 66 | |
BP315111.1 | - | 1 - 2 | 140 | 66 |