Assay Design



Gene Transcripts

Gene Symbol
NAT10
Entrez Gene ID
55226
Gene Name
N-acetyltransferase 10
Gene Aliases
ALP, NET43
Chromosome Location
Chr.11: 34105564 - 34146911 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144030.1 NP_001137502.1 1 - 2 229 81
GenBank mRNA AK294044.1 - 1 - 2 128 81

View Full Product Details