Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195214.1 | NP_001182143.1 | 4 - 5 | 584 | 80 |
NM_173834.3 | NP_776195.2 | 5 - 6 | 713 | 80 | |
XM_011530923.2 | XP_011529225.1 | 5 - 6 | 1121 | 80 | |
XM_011530924.1 | XP_011529226.1 | 5 - 6 | 495 | 80 | |
XM_017029450.1 | XP_016884939.1 | 5 - 6 | 495 | 80 | |
XM_017029451.1 | XP_016884940.1 | 4 - 5 | 992 | 80 | |
GenBank mRNA | AK292835.1 | - | 5 - 6 | 455 | 80 |
AK303992.1 | - | 4 - 5 | 314 | 80 | |
BC012469.1 | - | 5 - 6 | 451 | 80 |