Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282663.1 | NP_001269592.1 | 18 - 19 | 2635 | 58 |
NM_001282664.1 | NP_001269593.1 | 17 - 18 | 2488 | 58 | |
NM_014632.3 | NP_055447.1 | 18 - 19 | 2639 | 58 | |
XM_011520490.2 | XP_011518792.1 | 16 - 17 | 2342 | 58 | |
XM_011520491.2 | XP_011518793.1 | 16 - 17 | 2342 | 58 | |
XM_017018589.1 | XP_016874078.1 | 17 - 18 | 2455 | 58 | |
XM_017018590.1 | XP_016874079.1 | 17 - 18 | 2500 | 58 | |
XM_017018591.1 | XP_016874080.1 | 17 - 18 | 2455 | 58 | |
GenBank mRNA | AB018293.1 | - | 18 - 19 | 2600 | 58 |
AF052170.1 | - | 2 - 3 | 181 | 58 | |
AK294845.1 | - | 17 - 18 | 2455 | 58 | |
BC044577.1 | - | 18 - 19 | 2624 | 58 |