Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282663.1 | NP_001269592.1 | 25 - 26 | 3533 | 85 |
NM_001282664.1 | NP_001269593.1 | 23 - 24 | 3323 | 85 | |
NM_001282665.1 | NP_001269594.1 | 20 - 21 | 2816 | 85 | |
NM_001282666.1 | NP_001269595.1 | 18 - 19 | 2633 | 85 | |
NM_001282667.1 | NP_001269596.1 | 17 - 18 | 2570 | 85 | |
NM_014632.3 | NP_055447.1 | 25 - 26 | 3537 | 85 | |
XM_011520490.2 | XP_011518792.1 | 23 - 24 | 3240 | 85 | |
XM_011520491.2 | XP_011518793.1 | 23 - 24 | 3240 | 85 | |
XM_017018589.1 | XP_016874078.1 | 24 - 25 | 3353 | 85 | |
XM_017018590.1 | XP_016874079.1 | 24 - 25 | 3398 | 85 | |
XM_017018591.1 | XP_016874080.1 | 23 - 24 | 3290 | 85 | |
GenBank mRNA | AB018293.1 | - | 25 - 26 | 3498 | 85 |
AB110785.1 | - | 18 - 19 | 2556 | 85 | |
AB110786.1 | - | 17 - 18 | 2493 | 85 | |
AF052170.1 | - | 8 - 9 | 1016 | 85 | |
AK122821.1 | - | 3 - 4 | 3502 | 85 | |
AK127301.1 | - | 2 - 3 | 3830 | 85 | |
AK294845.1 | - | 23 - 24 | 85 | ||
AK302580.1 | - | 12 - 13 | 1425 | 85 | |
AK302893.1 | - | 21 - 22 | 3045 | 85 | |
BC044577.1 | - | 25 - 26 | 3522 | 85 | |
BX538021.1 | - | 15 - 16 | 2320 | 85 | |
BX641163.1 | - | 3 - 4 | 360 | 85 |