Assay Design



Gene Transcripts

Gene Symbol
FKTN
Entrez Gene ID
2218
Gene Name
fukutin
Gene Aliases
CMD1X, FCMD, LGMD2M, MDDGA4, MDDGB4, MDDGC4
Chromosome Location
Chr.9: 105558117 - 105655950 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079802.1 NP_001073270.1 6 - 7 863 124
NM_001198963.1 NP_001185892.1 6 - 7 863 124
NM_006731.2 NP_006722.2 5 - 6 771 124
XM_006717014.2 XP_006717077.1 5 - 6 775 124
XM_011518368.2 XP_011516670.1 7 - 8 1011 124
XM_011518369.2 XP_011516671.1 6 - 7 861 124
XM_011518370.2 XP_011516672.1 7 - 8 2547 124
XM_011518371.2 XP_011516673.1 7 - 8 1046 124
XM_011518373.2 XP_011516675.1 7 - 8 957 124
XM_011518374.2 XP_011516676.1 5 - 6 775 124
XM_011518375.2 XP_011516677.1 5 - 6 1090 124
XM_011518376.2 XP_011516678.1 5 - 6 913 124
XM_011518378.2 XP_011516680.1 7 - 8 1011 124
XM_011518379.2 XP_011516681.1 5 - 6 657 124
XM_011518381.2 XP_011516683.1 6 - 7 846 124
XM_011518387.2 XP_011516689.1 7 - 8 1011 124
XM_011518389.2 XP_011516691.1 7 - 8 1011 124
XM_011518390.2 XP_011516692.1 3 - 4 452 124
XM_011518391.2 XP_011516693.1 7 - 8 1011 124
XM_017014462.1 XP_016869951.1 8 - 9 1145 124
XM_017014463.1 XP_016869952.1 5 - 6 1234 124
XM_017014464.1 XP_016869953.1 7 - 8 1011 124
XM_017014465.1 XP_016869954.1 6 - 7 861 124
XM_017014466.1 XP_016869955.1 7 - 8 1011 124
XM_017014467.1 XP_016869956.1 6 - 7 1270 124
XM_017014468.1 XP_016869957.1 5 - 6 1153 124
XM_017014469.1 XP_016869958.1 7 - 8 1011 124
XM_017014470.1 XP_016869959.1 7 - 8 1011 124
XM_017014471.1 XP_016869960.1 8 - 9 1109 124
XM_017014472.1 XP_016869961.1 8 - 9 1135 124
XM_017014473.1 XP_016869962.1 3 - 4 3013 124
XM_017014474.1 XP_016869963.1 5 - 6 774 124
XM_017014475.1 XP_016869964.1 5 - 6 778 124
XM_017014476.1 XP_016869965.1 8 - 9 1129 124
XM_017014477.1 XP_016869966.1 7 - 8 994 124
XM_017014478.1 XP_016869967.1 9 - 10 1196 124
XM_017014479.1 XP_016869968.1 6 - 7 893 124
GenBank mRNA AB008226.1 - 5 - 6 758 124
AB609007.1 - - 758 124
AK300840.1 - 6 - 7 861 124
AK304460.1 - 5 - 6 769 124
BC101808.1 - 4 - 5 673 124
BC112038.1 - 4 - 5 673 124
BC117699.1 - 4 - 5 679 124
BC117700.1 - 6 - 7 921 124

View Full Product Details