Assay Design



Gene Transcripts

Gene Symbol
ARNT
Entrez Gene ID
405
Gene Name
aryl hydrocarbon receptor nuclear translocator
Gene Aliases
HIF-1-beta, HIF-1beta, HIF1-beta, HIF1B, HIF1BETA, TANGO, bHLHe2
Chromosome Location
Chr.1: 150809705 - 150876768 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197325.1 NP_001184254.1 15 - 16 1736 61
NM_001286035.1 NP_001272964.1 17 - 18 1822 61
NM_001286036.1 NP_001272965.1 16 - 17 1781 61
NM_001668.3 NP_001659.1 16 - 17 1781 61
NM_178427.2 NP_848514.1 15 - 16 1736 61
XM_005245151.1 XP_005245208.1 16 - 17 1723 61
XM_005245153.1 XP_005245210.1 16 - 17 1708 61
XM_005245154.3 XP_005245211.1 17 - 18 1751 61
XM_005245157.1 XP_005245214.1 15 - 16 1612 61
XM_011509542.2 XP_011507844.1 16 - 17 1784 61
XM_011509543.2 XP_011507845.1 17 - 18 1779 61
XM_011509544.2 XP_011507846.1 16 - 17 1748 61
XM_011509545.2 XP_011507847.1 15 - 16 1739 61
XM_011509546.1 XP_011507848.1 15 - 16 1499 61
XM_011509547.2 XP_011507849.1 15 - 16 1739 61
XM_017001288.1 XP_016856777.1 17 - 18 1626 61
XM_017001289.1 XP_016856778.1 17 - 18 1611 61
XM_017001290.1 XP_016856779.1 16 - 17 1580 61
XM_017001291.1 XP_016856780.1 16 - 17 1582 61
XM_017001292.1 XP_016856781.1 16 - 17 1516 61
XM_017001293.1 XP_016856782.1 15 - 16 1473 61
XM_017001294.1 XP_016856783.1 15 - 16 1612 61
XM_017001295.1 XP_016856784.1 15 - 16 1597 61
XM_017001296.1 XP_016856785.1 14 - 15 1576 61
GenBank mRNA AB209877.1 - 16 - 17 1592 61
AK290177.1 - 16 - 17 1720 61
AK291705.1 - 15 - 16 1578 61
AK293027.1 - 16 - 17 1781 61
AK304772.1 - 14 - 15 1474 61
AK313780.1 - 16 - 17 1722 61
AK315812.1 - 15 - 16 1575 61
AL834279.1 - 17 - 18 1761 61
BC041121.1 - 16 - 17 1595 61
M69238.1 - 16 - 17 1636 61

View Full Product Details